Assay Details
| Gene Symbol: Car14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011797.2 | NP_035927.1 | 4 - 5 | 668 | 85 | |
| GenBank mRNA | AB005450.1 | - | 4 - 5 | 668 | 85 | |
| AK157764.1 | - | 4 - 5 | 668 | 85 | ||
| BC046995.1 | - | 4 - 5 | 650 | 6491779 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car14
Genomic Map