Assay Details
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011842.3 | NP_035972.3 | 16 - 17 | 2127 | 72 | |
GenBank mRNA | AF159259.1 | - | 16 - 17 | 2002 | 72 | |
AK003701.1 | - | 4 - 5 | 561 | 72 | ||
AK088158.1 | - | 16 - 17 | 2040 | 72 | ||
AK147099.1 | - | 16 - 17 | 2106 | 72 | ||
AK157717.1 | - | 16 - 17 | 2031 | 72 | ||
AK159979.1 | - | 16 - 17 | 2103 | 72 | ||
AK198824.1 | - | 2 - 3 | 153 | 72 | ||
BC079847.1 | - | 16 - 17 | 2127 | 6419411 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
