Assay Details
| Gene Symbol: Ctsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012007.1 | NP_036137.1 | 3 - 4 | 322 | 128 | |
| GenBank mRNA | AF136272.1 | - | 3 - 4 | 322 | 128 | |
| AF158182.1 | - | 2 - 3 | 262 | 128 | ||
| AK005526.1 | - | 3 - 4 | 329 | 128 | ||
| AK131661.1 | - | 3 - 4 | 329 | 128 | ||
| BC103769.1 | - | 3 - 4 | 318 | 30787117 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002898 | AK005526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002898 | BC103769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002898 | NM_012007.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002898 | AF136272.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747931 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002898 | AK005526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002898 | BC103769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002898 | NM_012007.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002898 | AF136272.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427282 | NM_012007.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427282 | AF136272.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427282 | AK005526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427282 | AK131661.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427282 | AF158182.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177472 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02706945 | - | - |
| Whole Mouse Genome G4122A | A_51_P427282 | NM_012007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427282 | AF136272.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427282 | AK005526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427282 | AK131661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427282 | AF158182.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101401 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10555916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10975756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10927724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10786028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10786007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11027173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10969915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10831518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10975768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29088288 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409890 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416413_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409887 | - | - |
| Mouse Genome 430 2.0 Array | 1416413_at | - | - |
| Mouse Genome 430 2.0 Array | 1416413_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26898_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26898_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013772.mm.1 | NM_012007.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013772.mm.1 | NM_012007.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013772.mm.1 | TC1300002145.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416413_PM_at | - | - |
| Mouse Expression Set 430 | 1416413_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416413_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416413_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416413_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416413_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 771425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1660398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228474 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228474 | - | - |
| mouse-6 v1.1 (Illumina) | scl026898.1_219-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026898.1_219-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228474 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228474 | - | - |

Ctsj
Genomic Map