Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_001253897.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_001253897.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_001253898.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_001253898.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_012029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
NM_012029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016004.mm.1 |
TC0900001898.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_001253897.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_001253897.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_001253898.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_001253898.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_012029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
NM_012029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016005.mm.1 |
TC0900001898.mm.1 |
MTA TC |