Assay Details
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083960.1 | NP_001077429.1 | 3 - 4 | 340 | 101 | |
NM_012046.2 | NP_036176.1 | 4 - 5 | 454 | 101 | ||
GenBank mRNA | AF149309.1 | - | 3 - 4 | 303 | 101 | |
AF163053.1 | - | 3 - 4 | 340 | 101 | ||
AF163054.1 | - | 3 - 4 | 340 | 101 | ||
AF165313.1 | - | 3 - 4 | 333 | 101 | ||
AF169386.1 | - | 4 - 5 | 451 | 101 | ||
AF173848.1 | - | 4 - 5 | 447 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
