Assay Details
Gene Symbol: Msh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF107352.1 | - | 10 - 11 | 848 | 106 | |
AF146227.1 | - | 9 - 10 | 1056 | 106 | ||
BC141113.1 | - | 9 - 10 | 976 | 9055725 | 106 | |
BC145406.1 | - | 9 - 10 | 976 | 106 | ||
BC145407.1 | - | 9 - 10 | 976 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
