Assay Details
| Gene Symbol: Surf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271724.1 | NP_001258653.2 | 2 - 3 | 324 | 56 | |
| NM_013677.2 | NP_038705.2 | 2 - 3 | 324 | 56 | ||
| GenBank mRNA | AK002469.1 | - | 2 - 3 | 138 | 56 | |
| AK154726.1 | - | 2 - 3 | 144 | 56 | ||
| BC004755.1 | - | 2 - 3 | 324 | 3584373 | 56 | |
| BU697260.1 | - | 2 - 3 | 277 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Surf1
Genomic Map