Assay Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253877.1 | NP_001240806.1 | 11 - 12 | 1641 | 89 | |
NM_001253878.1 | NP_001240807.1 | 11 - 12 | 1360 | 89 | ||
NM_001253879.1 | NP_001240808.1 | 10 - 11 | 1283 | 89 | ||
NM_001253880.1 | NP_001240809.1 | 8 - 9 | 998 | 89 | ||
NM_013827.3 | NP_038855.2 | 10 - 11 | 1279 | 89 | ||
NR_045608.1 | 12 - 13 | 1347 | 89 | |||
GenBank mRNA | AK030752.1 | - | 10 - 11 | 1229 | 89 | |
AK031409.1 | - | 12 - 13 | 1348 | 89 | ||
AK032798.1 | - | 11 - 12 | 1602 | 89 | ||
AK032819.1 | - | 11 - 12 | 1603 | 89 | ||
AK137539.1 | - | 12 - 13 | 1588 | 89 | ||
AK139685.1 | - | 11 - 12 | 1361 | 89 | ||
BC050826.1 | - | 10 - 11 | 1233 | 89 | ||
BC057163.1 | - | 10 - 11 | 1076 | 89 | ||
BC071223.1 | - | 8 - 9 | 751 | 89 | ||
BC092237.1 | - | 10 - 11 | 1211 | 30637248 | 89 | |
BC100340.1 | - | 8 - 9 | 929 | 30715034 | 89 | |
S78454.1 | - | 7 - 8 | 683 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
