Assay Details
| Gene Symbol: Cabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013877.3 | NP_038905.1 | 1 - 2 | 211 | 81 | |
| GenBank mRNA | AF169161.1 | - | 1 - 2 | 172 | 81 | |
| AK044544.1 | - | 1 - 2 | 212 | 81 | ||
| AK087416.1 | - | 1 - 2 | 72 | 81 | ||
| BC018253.1 | - | 1 - 2 | 234 | 4504283 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055504 | AK087416.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055504 | BC018253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055504 | NM_013877.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055504 | AB079605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055504 | AF169161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033215 | BC018253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033215 | NM_013877.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033215 | AK044544.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146721 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123378 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107746 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033215 | BC018253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033215 | NM_013877.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033215 | AK044544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055504 | AK087416.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055504 | BC018253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055504 | NM_013877.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055504 | AB079605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055504 | AF169161.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24975623 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123378 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107746 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06126650 | - | - |
| Whole Mouse Genome G4122A | A_51_P354289 | BC018253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354289 | NM_013877.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354289 | AF169161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354289 | AK044544.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110508 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103545 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06629114 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06629112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06629109 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06127525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06629115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24975653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06347518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06489308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06555307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06212466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06555306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06085913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06629109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06112007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06456543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05940913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06640040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24975629 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550154 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451826_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421780_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550146 | - | - |
| Mouse Genome 430 2.0 Array | 1421780_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421780_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451826_at | - | - |
| Mouse Genome 430 2.0 Array | 1451826_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 29865_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 29865_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700001319.mm.1 | TC0700000165.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001319.mm.1 | NM_013877.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001319.mm.1 | NM_013877.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001322.mm.1 | TC0700000165.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001322.mm.1 | NM_013877.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001322.mm.1 | NM_013877.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001323.mm.1 | TC0700000165.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001323.mm.1 | NM_013877.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001323.mm.1 | NM_013877.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001321.mm.1 | TC0700000165.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001321.mm.1 | NM_013877.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001321.mm.1 | NM_013877.3 | Refseq |
| Mouse Expression Set 430 | 1451826_PM_at | - | - |
| Mouse Expression Set 430 | 1421780_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451826_at | - | - |
| Mouse Expression Set 430 | 1421780_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451826_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421780_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421780_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451826_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 444761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106760672 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237612 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237612 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217946 | - | - |
| mouse-6 v1.1 (Illumina) | scl33068.6_244-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360273 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106760672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33068.6_244-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106760672 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217946 | - | - |

Cabp5
Genomic Map