Assay Details
| Gene Symbol: Col5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015734.2 | NP_056549.2 | 13 - 14 | 2057 | 67 | |
| GenBank mRNA | AB009993.1 | - | 13 - 14 | 1797 | 67 | |
| BC138447.1 | - | 13 - 14 | 1731 | 8861467 | 67 | |
| BC138449.1 | - | 13 - 14 | 1731 | 8861469 | 67 | |
| BQ885116.1 | - | 1 - 2 | 51 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col5a1
Genomic Map