Assay Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015747.2 | NP_056562.1 | 10 - 11 | 2379 | 57 | |
| GenBank mRNA | AK155506.1 | - | 10 - 11 | 2352 | 57 | |
| BC015085.1 | - | 10 - 11 | 2279 | 4015575 | 57 | |
| M73696.1 | - | 9 - 10 | 2298 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a1
Genomic Map