Assay Details
| Gene Symbol: Mfap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015776.2 | NP_056591.1 | 9 - 10 | 444 | 83 | |
| GenBank mRNA | AF180805.1 | - | 9 - 10 | 444 | 83 | |
| AK003479.1 | - | 9 - 10 | 461 | 83 | ||
| AK011458.1 | - | 9 - 10 | 521 | 83 | ||
| BC041146.1 | - | 3 - 4 | 2176 | 83 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062777 | BC041146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062777 | NM_015776.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062777 | AF180805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062777 | AK003479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062777 | AK011458.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825986 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66838 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137824 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153774 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062777 | BC041146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062777 | NM_015776.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062777 | AF180805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062777 | AK003479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062777 | AK011458.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363681 | NM_015776.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363681 | AF180805.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363681 | AK003479.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24809078 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137824 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153774 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06052423 | - | - |
| Whole Mouse Genome G4122A | A_51_P363681 | NM_015776.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363681 | AF180805.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363681 | AK003479.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115637 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153774 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05743309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05091660 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05743307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05181084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05334965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24809093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05172245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05007880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05421374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05266394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05089181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05197943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05421373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05175906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05181084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05421369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05797880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05011927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05787482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05638451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05691065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05536823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05840672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05089182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05175910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05743308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05683690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05011929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05089186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05216409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05175905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05840671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05724359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05536821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05011924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05421370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24809221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05840666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05691064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10541497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541501 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449082_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418454_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10541496 | - | - |
| Mouse Genome 430 2.0 Array | 1449082_at | - | - |
| Mouse Genome 430 2.0 Array | 1449082_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418454_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418454_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50530_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50530_at | - | - |
| Mouse Expression Set 430 | 1418454_PM_at | - | - |
| Mouse Expression Set 430 | 1418454_at | - | - |
| Mouse Expression Set 430 | 1449082_PM_at | - | - |
| Mouse Expression Set 430 | 1449082_at | - | - |
| Murine Genome U74v2 Set | 99517_at | - | - |
| Murine Genome U74v2 Set | 99518_at | - | - |
| Murine Genome U74v2 Set | 99517_PM_at | - | - |
| Murine Genome U74v2 Set | 99518_PM_at | - | - |
| Murine Genome U74v2 Set | 169062_at | - | - |
| Murine Genome U74v2 Set | 131248_PM_at | - | - |
| Murine Genome U74v2 Set | 169062_PM_at | - | - |
| Murine Genome U74v2 Set | 131248_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418454_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449082_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418454_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449082_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418454_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449082_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449082_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418454_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 625696 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225835 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225835 | - | - |
| mouse-6 v1.1 (Illumina) | scl29544.11_278-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29544.11_278-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225835 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225835 | - | - |

Mfap5
Genomic Map