Assay Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015830.1 | NP_056645.1 | 7 - 8 | 2536 | 65 | |
| GenBank mRNA | AK079422.1 | - | 6 - 7 | 2628 | 65 | |
| AK143711.1 | - | 9 - 10 | 3175 | 65 | ||
| AK148209.1 | - | 5 - 6 | 795 | 65 | ||
| AK170543.1 | - | 8 - 9 | 2770 | 65 | ||
| AK181088.1 | - | 1 - 2 | 80 | 65 | ||
| BC058094.1 | - | 8 - 9 | 2794 | 6824892 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn15
Genomic Map