Assay Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016695.3 | NP_057904.1 | 8 - 9 | 1047 | 99 | |
| GenBank mRNA | AF162685.1 | - | 8 - 9 | 1034 | 99 | |
| AK161577.1 | - | 8 - 9 | 1040 | 99 | ||
| AK172253.1 | - | 8 - 9 | 1032 | 99 | ||
| BC053026.1 | - | 8 - 9 | 1047 | 5709543 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp2
Genomic Map