Assay Details
| Gene Symbol: Cd164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016898.2 | NP_058594.1 | 1 - 2 | 199 | 80 | |
| GenBank mRNA | AB014464.1 | - | 1 - 2 | 164 | 80 | |
| AB028895.1 | - | 1 - 2 | 199 | 80 | ||
| AF299345.1 | - | 1 - 2 | 164 | 80 | ||
| AK018009.1 | - | 1 - 2 | 199 | 80 | ||
| AK145131.1 | - | 1 - 2 | 258 | 80 | ||
| AK169275.1 | - | 1 - 2 | 239 | 80 | ||
| BC005414.1 | - | 1 - 2 | 187 | 3600535 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd164
Genomic Map