Assay Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016906.4 | NP_058602.1 | 4 - 5 | 331 | 68 | |
| GenBank mRNA | AB032902.1 | - | 4 - 5 | 343 | 68 | |
| AF145253.1 | - | 4 - 5 | 253 | 68 | ||
| AF222743.1 | - | 4 - 5 | 253 | 68 | ||
| AK088473.1 | - | 4 - 5 | 332 | 68 | ||
| AK153055.1 | - | 4 - 5 | 321 | 68 | ||
| AK159349.1 | - | 4 - 5 | 319 | 68 | ||
| AK167484.1 | - | 4 - 5 | 261 | 68 | ||
| AK219834.1 | - | 3 - 4 | 174 | 68 | ||
| BC003707.1 | - | 4 - 5 | 257 | 3592772 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a1
Genomic Map