Assay Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016906.4 | NP_058602.1 | 6 - 7 | 569 | 96 | |
| GenBank mRNA | AB032902.1 | - | 6 - 7 | 581 | 96 | |
| AF145253.1 | - | 6 - 7 | 491 | 96 | ||
| AF222743.1 | - | 6 - 7 | 491 | 96 | ||
| AK054318.1 | - | 1 - 2 | 1405 | 96 | ||
| AK088473.1 | - | 6 - 7 | 570 | 96 | ||
| AK153055.1 | - | 6 - 7 | 559 | 96 | ||
| AK159349.1 | - | 6 - 7 | 557 | 96 | ||
| AK167484.1 | - | 6 - 7 | 499 | 96 | ||
| AK197110.1 | - | 2 - 3 | 181 | 96 | ||
| AK198895.1 | - | 2 - 3 | 201 | 97 | ||
| BC003707.1 | - | 6 - 7 | 495 | 3592772 | 96 | |
| BC006020.1 | - | 2 - 3 | 154 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a1
Genomic Map