Assay Details
| Gene Symbol: Clca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017474.2 | NP_059502.1 | 2 - 3 | 334 | 64 | |
| GenBank mRNA | AB017156.1 | - | 2 - 3 | 317 | 64 | |
| AK007466.1 | - | 2 - 3 | 331 | 64 | ||
| AK008659.2 | - | 2 - 3 | 331 | 64 | ||
| AK033560.1 | - | 335 | 64 | |||
| AK136476.1 | - | 2 - 3 | 331 | 64 | ||
| BC028343.1 | - | 2 - 3 | 324 | 4217548 | 64 | |
| BC116318.2 | - | 2 - 3 | 317 | 40095991 | 64 | |
| BC116319.2 | - | 2 - 3 | 317 | 40095992 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clca1
Genomic Map