Assay Details
| Gene Symbol: Elp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253700.1 | NP_001240629.1 | 1 - 2 | 124 | 92 | |
| NM_018740.2 | NP_061210.2 | 2 - 3 | 385 | 92 | ||
| GenBank mRNA | AA646690.1 | - | 1 - 2 | 40 | 92 | |
| AF004107.1 | - | 1 - 2 | 304 | 92 | ||
| AK152582.1 | - | 2 - 3 | 389 | 92 | ||
| BC003752.1 | - | 1 - 2 | 236 | 3489878 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253700.1 transcripts.
Set Membership: |
|

Elp5
Genomic Map