Assay Details
| Gene Symbol: Elp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018740.2 | NP_061210.2 | 5 - 6 | 742 | 95 | |
| GenBank mRNA | AF004107.1 | - | 4 - 5 | 661 | 95 | |
| AK011130.1 | - | 3 - 4 | 554 | 95 | ||
| AK013662.1 | - | 3 - 4 | 343 | 95 | ||
| AK152582.1 | - | 5 - 6 | 746 | 95 | ||
| AK199980.1 | - | 2 - 3 | 288 | 95 | ||
| AK207946.1 | - | 1 - 2 | 224 | 95 | ||
| BC003752.1 | - | 4 - 5 | 593 | 3489878 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elp5
Genomic Map