Assay Details
| Gene Symbol: Smarcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018817.2 | NP_061287.2 | 7 - 8 | 1609 | 75 | |
| GenBank mRNA | AF088884.1 | - | 7 - 8 | 1589 | 75 | |
| AK042332.1 | - | 7 - 8 | 1588 | 75 | ||
| AK077878.1 | - | 7 - 8 | 1587 | 75 | ||
| AK083488.1 | - | 7 - 8 | 1587 | 75 | ||
| AK154020.1 | - | 7 - 8 | 1592 | 75 | ||
| AK161570.1 | - | 7 - 8 | 1529 | 75 | ||
| BC029078.1 | - | 7 - 8 | 1544 | 3992325 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcal1
Genomic Map