Assay Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018884.2 | NP_061372.2 | 4 - 5 | 1176 | 108 | |
| GenBank mRNA | AF127084.1 | - | 4 - 5 | 1176 | 108 | |
| AF127085.1 | - | 5 - 6 | 1020 | 108 | ||
| AK143226.1 | - | 4 - 5 | 690 | 108 | ||
| AK173098.1 | - | 4 - 5 | 1135 | 108 | ||
| HM560981.1 | - | 4 - 5 | 1167 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdzrn3
Genomic Map