Assay Details
| Gene Symbol: Vcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081249.1 | NP_001074718.1 | 9 - 10 | 9695 | 72 | |
| NM_001134474.1 | NP_001127946.1 | 8 - 9 | 4478 | 72 | ||
| NM_001134475.1 | NP_001127947.1 | 7 - 8 | 1598 | 72 | ||
| NM_019389.2 | NP_062262.2 | 8 - 9 | 6815 | 72 | ||
| GenBank mRNA | AK045314.1 | - | 2 - 3 | 3150 | 72 | |
| D16263.1 | - | 7 - 8 | 6387 | 72 | ||
| D28599.1 | - | 9 - 10 | 9439 | 72 | ||
| D32040.1 | - | 7 - 8 | 1330 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vcan
Genomic Map