Assay Details
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205311.1 | NP_001192240.1 | 2 - 3 | 451 | 57 | |
NM_019415.2 | NP_062288.2 | 2 - 3 | 448 | 57 | ||
GenBank mRNA | AI882166.1 | - | 2 - 3 | 440 | 57 | |
AK052691.1 | - | 2 - 3 | 452 | 57 | ||
AK052755.1 | - | 2 - 3 | 450 | 57 | ||
AK085496.1 | - | 2 - 3 | 455 | 57 | ||
BC038612.1 | - | 2 - 3 | 473 | 4237274 | 57 | |
U61085.1 | - | 2 - 3 | 430 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
