Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 7 - 8 | 1128 | 74 | |
| GenBank mRNA | AF127656.1 | - | 8 - 9 | 976 | 74 | |
| AF127657.1 | - | 7 - 8 | 919 | 74 | ||
| AF127658.1 | - | 7 - 8 | 1128 | 74 | ||
| AF127659.1 | - | 7 - 8 | 1128 | 74 | ||
| AF127660.1 | - | 8 - 9 | 976 | 74 | ||
| AF127661.1 | - | 7 - 8 | 919 | 74 | ||
| AF127662.1 | - | 8 - 9 | 1047 | 74 | ||
| AK002625.1 | - | 8 - 9 | 910 | 74 | ||
| AK146917.1 | - | 8 - 9 | 999 | 74 | ||
| BC012214.1 | - | 8 - 9 | 981 | 3967218 | 74 | |
More Information
Set Membership: |
|

Rabggta
Genomic Map