Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 14 - 15 | 1795 | 115 | |
| GenBank mRNA | AF127656.1 | - | 15 - 16 | 1643 | 115 | |
| AF127657.1 | - | 14 - 15 | 1586 | 115 | ||
| AF127658.1 | - | 14 - 15 | 1795 | 115 | ||
| AF127659.1 | - | 14 - 15 | 1795 | 115 | ||
| AF127660.1 | - | 14 - 15 | 1740 | 115 | ||
| AF127661.1 | - | 13 - 14 | 1683 | 115 | ||
| AF127662.1 | - | 15 - 16 | 1714 | 115 | ||
| AK002625.1 | - | 15 - 16 | 1577 | 115 | ||
| AK146917.1 | - | 15 - 16 | 1666 | 115 | ||
| BC012214.1 | - | 13 - 14 | 1829 | 3967218 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabggta
Genomic Map