Assay Details
| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019521.2 | NP_062394.2 | 7 - 8 | 902 | 96 | |
| GenBank mRNA | AK086187.1 | - | 7 - 8 | 897 | 96 | |
| BC005444.1 | - | 7 - 8 | 857 | 3592398 | 96 | |
| X59846.1 | - | 7 - 8 | 873 | 96 | ||
More Information
Set Membership: |
|

Gas6
Genomic Map