Assay Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019667.2 | NP_062641.1 | 5 - 6 | 494 | 119 | |
| GenBank mRNA | AB012611.1 | - | 5 - 6 | 509 | 119 | |
| AK004604.2 | - | 5 - 6 | 494 | 119 | ||
| AK035789.1 | - | 547 | 119 | |||
| AK044230.1 | - | 4 - 5 | 423 | 119 | ||
| AK078244.1 | - | 5 - 6 | 526 | 119 | ||
| AK159699.1 | - | 5 - 6 | 523 | 119 | ||
| AK168729.1 | - | 5 - 6 | 526 | 119 | ||
| BC013818.1 | - | 5 - 6 | 530 | 3671426 | 119 | |
| DQ141211.1 | - | 6 - 7 | 724 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stam2
Genomic Map