Assay Details
| Gene Symbol: Mmp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019724.3 | NP_062698.2 | 5 - 6 | 1112 | 95 | |
| GenBank mRNA | AB021228.1 | - | 5 - 6 | 927 | 95 | |
| AF282844.1 | - | 5 - 6 | 879 | 95 | ||
| AK048543.1 | - | 5 - 6 | 992 | 95 | ||
| AK077939.1 | - | 1155 | 95 | |||
| BC057926.1 | - | 5 - 6 | 1114 | 5292478 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp16
Genomic Map