Assay Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019724.3 | NP_062698.2 | 6 - 7 | 1323 | 86 | |
GenBank mRNA | AB021228.1 | - | 6 - 7 | 1138 | 86 | |
AF282844.1 | - | 6 - 7 | 1090 | 86 | ||
AK048543.1 | - | 6 - 7 | 1203 | 86 | ||
AK077939.1 | - | 1366 | 86 | |||
BC057926.1 | - | 6 - 7 | 1325 | 5292478 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
