Assay Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019789.4 | NP_062763.2 | 2 - 3 | 317 | 88 | |
| GenBank mRNA | AF184624.1 | - | 2 - 3 | 265 | 88 | |
| AF274050.1 | - | 1 - 2 | 177 | 88 | ||
| AF287733.1 | - | 2 - 3 | 295 | 88 | ||
| AF300870.1 | - | 2 - 3 | 240 | 88 | ||
| AK165711.1 | - | 2 - 3 | 265 | 88 | ||
| BC026980.1 | - | 2 - 3 | 254 | 5368268 | 88 | |
| BC047139.1 | - | 2 - 3 | 254 | 6389719 | 88 | |
| BC057329.1 | - | 2 - 3 | 236 | 6813819 | 88 | |
| DQ148499.1 | - | 1 - 2 | 177 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnip3
Genomic Map