Assay Details
| Gene Symbol: Stat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019963.1 | NP_064347.1 | 18 - 19 | 1777 | 75 | |
| GenBank mRNA | AF088862.1 | - | 18 - 19 | 1650 | 75 | |
| AF187231.1 | - | 18 - 19 | 1774 | 75 | ||
| AF206162.1 | - | 18 - 19 | 1777 | 75 | ||
| AK030903.1 | - | 11 - 12 | 869 | 75 | ||
| AK149924.1 | - | 18 - 19 | 1790 | 75 | ||
| AK150070.1 | - | 18 - 19 | 1789 | 75 | ||
| AK153405.1 | - | 16 - 17 | 1424 | 75 | ||
| BC064827.1 | - | 7 - 8 | 774 | 75 | ||
More Information
Set Membership: |
|

Stat2
Genomic Map