Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600007816.mm.1 |
NM_019973.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007816.mm.1 |
NM_019973.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007816.mm.1 |
NM_178880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007816.mm.1 |
NM_178880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007816.mm.1 |
TC1600001008.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007815.mm.1 |
NM_019973.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007815.mm.1 |
NM_019973.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007815.mm.1 |
NM_178880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007815.mm.1 |
NM_178880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007815.mm.1 |
TC1600001008.mm.1 |
MTA TC |