Assay Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020260.2 | NP_064656.2 | 2 - 3 | 569 | 70 | |
| GenBank mRNA | AF151363.1 | - | 2 - 3 | 210 | 70 | |
| AK018057.1 | - | 2 - 3 | 435 | 70 | ||
| AK138417.1 | - | 2 - 3 | 809 | 70 | ||
| AK149975.1 | - | 2 - 3 | 405 | 70 | ||
| AK150109.1 | - | 2 - 3 | 405 | 70 | ||
| AK155729.1 | - | 2 - 3 | 389 | 70 | ||
| BB865916.1 | - | 2 - 3 | 571 | 70 | ||
| BC138870.1 | - | 2 - 3 | 374 | 8861892 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap31
Genomic Map