Assay Details
| Gene Symbol: Vsig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020518.2 | NP_065264.2 | 6 - 7 | 974 | 137 | |
| GenBank mRNA | AK004478.1 | - | 5 - 6 | 694 | 137 | |
| BC099457.1 | - | 4 - 5 | 659 | 30789633 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160482 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083879 | NM_020518.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083879 | AK004478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083879 | BC099457.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9691 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174070 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083879 | NM_020518.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083879 | AK004478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083879 | BC099457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160482 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431198 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174070 | - | - |
| Whole Mouse Genome G4122A | A_52_P391743 | NM_020518.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391743 | AK004478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391743 | AK008920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391743 | BC099457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160482 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113380 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100788 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07596632 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07596638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07596637 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07764696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07596628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07596627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08081537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08022885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08022891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07776148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07776142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08076054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431227 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584331 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422634_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584325 | - | - |
| Mouse Genome 430 2.0 Array | 1422634_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422634_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57276_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57276_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900002736.mm.1 | NM_020518.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002736.mm.1 | NM_020518.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002736.mm.1 | TC0900000369.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422634_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422634_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422634_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422634_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422634_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422634_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 539557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6420075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245518 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2762841 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2762841 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245518 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003483.1_26-S | - | - |
| mouse-6 v1.1 (Illumina) | scl37111.7.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720609 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37111.7.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003483.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2762841 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245518 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2762841 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245518 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003483.1_26-S | - | - |

Vsig2
Genomic Map