Assay Details
| Gene Symbol: Smpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021491.3 | NP_067466.1 | 5 - 6 | 2018 | 143 | |
| GenBank mRNA | AJ250461.1 | - | 3 - 4 | 1549 | 143 | |
| AK019476.2 | - | 6 - 7 | 2070 | 143 | ||
| AK076416.1 | - | 5 - 6 | 1995 | 143 | ||
| AK139226.1 | - | 5 - 6 | 1994 | 143 | ||
| BC043077.1 | - | 5 - 6 | 2018 | 5721118 | 143 | |
| BC046980.1 | - | 4 - 5 | 1838 | 6399438 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd3
Genomic Map