Assay Details
| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022012.3 | NP_071295.2 | 8 - 9 | 2336 | 62 | |
| GenBank mRNA | AK159518.1 | - | 6 - 7 | 1356 | 62 | |
| BC030928.1 | - | 6 - 7 | 889 | 62 | ||
| BC047152.1 | - | 6 - 7 | 2083 | 6476975 | 62 | |
| BC095963.1 | - | 8 - 9 | 2336 | 5400993 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map3k11
Genomic Map