Assay Details
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022317.3 | NP_071712.3 | 13 - 14 | 1561 | 77 | |
GenBank mRNA | AF305211.1 | - | 13 - 14 | 1494 | 77 | |
AK052784.1 | - | 13 - 14 | 1562 | 77 | ||
AK145077.1 | - | 13 - 14 | 1562 | 77 | ||
BC010472.1 | - | 13 - 14 | 1549 | 3586005 | 77 | |
BC013783.1 | - | 13 - 14 | 1552 | 3600786 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
