| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
TC0600000135.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001083315.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001289624.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001289625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001289626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001289627.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_001289629.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000966.mm.1 |
NM_022332.3 |
Refseq |