Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900014916.mm.1 |
NM_022888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014916.mm.1 |
NM_022888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014916.mm.1 |
NM_176807.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014916.mm.1 |
NM_176807.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014916.mm.1 |
TC0900001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014918.mm.1 |
NM_022888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014918.mm.1 |
NM_022888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014918.mm.1 |
NM_176807.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014918.mm.1 |
NM_176807.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014918.mm.1 |
TC0900001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014920.mm.1 |
NM_022888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014920.mm.1 |
NM_022888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014920.mm.1 |
NM_176807.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014920.mm.1 |
NM_176807.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014920.mm.1 |
TC0900001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014921.mm.1 |
NM_022888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014921.mm.1 |
NM_022888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014921.mm.1 |
NM_176807.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014921.mm.1 |
NM_176807.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014921.mm.1 |
TC0900001779.mm.1 |
MTA TC |