Assay Details
| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023044.2 | NP_075531.2 | 5 - 6 | 1744 | 68 | |
| GenBank mRNA | AF121080.1 | - | 5 - 6 | 1752 | 68 | |
| AK051941.1 | - | 5 - 6 | 1345 | 68 | ||
| AK171698.1 | - | 1397 | 68 | |||
| BC051940.1 | - | 5 - 6 | 1415 | 5717926 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a3
Genomic Map