Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015315.mm.1 |
NM_001164704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015315.mm.1 |
NM_001164704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015315.mm.1 |
NM_023132.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015315.mm.1 |
NM_023132.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015315.mm.1 |
TC0X00002440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015323.mm.1 |
NM_001164704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015323.mm.1 |
NM_001164704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015323.mm.1 |
NM_023132.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015323.mm.1 |
NM_023132.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015323.mm.1 |
TC0X00002440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015321.mm.1 |
NM_001164704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015321.mm.1 |
NM_001164704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015321.mm.1 |
NM_023132.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015321.mm.1 |
NM_023132.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015321.mm.1 |
TC0X00002440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015317.mm.1 |
NM_001164704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015317.mm.1 |
NM_001164704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015317.mm.1 |
NM_023132.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015317.mm.1 |
NM_023132.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015317.mm.1 |
TC0X00002440.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015324.mm.1 |
NM_001164704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015324.mm.1 |
NM_001164704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015324.mm.1 |
NM_023132.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015324.mm.1 |
NM_023132.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015324.mm.1 |
TC0X00002440.mm.1 |
MTA TC |