Assay Details
| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025656.4 | NP_079932.2 | 8 - 9 | 797 | 83 | |
| GenBank mRNA | AK005928.1 | - | 8 - 9 | 83 | ||
| AK007515.1 | - | 8 - 9 | 798 | 83 | ||
| AK013414.1 | - | 8 - 9 | 754 | 83 | ||
| BC053424.1 | - | 8 - 9 | 1226 | 6336522 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469866 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107830 | AK013414.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107830 | BC053424.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107830 | NM_025656.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107830 | AK005928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107830 | AK007515.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112599 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173861 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129267 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107830 | AK013414.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107830 | BC053424.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107830 | NM_025656.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107830 | AK005928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107830 | AK007515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315101 | AK013414.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315101 | BC053424.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315101 | AK005928.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28469870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07635079 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02526095 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129267 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07635093 | - | - |
| Whole Mouse Genome G4122A | A_52_P544666 | AK013414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544666 | BC053424.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544666 | NM_025656.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544666 | AK005928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544666 | AK007515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315101 | AK013414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315101 | BC053424.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315101 | AK005928.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104526 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137446 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129267 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07635093 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10194052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10194043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10194049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10194050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28469866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28469856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10041677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10160872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10194041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10104788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09748639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10045201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09748644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09784400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28469879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395897 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451044_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458627_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395893 | - | - |
| Mouse Genome 430 2.0 Array | 1451044_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458627_at | - | - |
| Mouse Genome 430 2.0 Array | 1451044_at | - | - |
| Mouse Genome 430 2.0 Array | 1458627_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66603_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66603_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003459.mm.1 | NM_025656.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003459.mm.1 | TC1200000498.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451044_at | - | - |
| Mouse Expression Set 430 | 1451044_PM_at | - | - |
| Mouse Expression Set 430 | 1458627_PM_at | - | - |
| Mouse Expression Set 430 | 1458627_at | - | - |
| Murine Genome U74v2 Set | 166221_r_at | - | - |
| Murine Genome U74v2 Set | 166221_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451044_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451044_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451044_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451044_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458627_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458627_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 414755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2640706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255390 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255390 | - | - |
| mouse-6 v1.1 (Illumina) | scl43183.10.1_88-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43183.10.1_88-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2920800 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2920800 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255390 | - | - |

Gemin2
Genomic Map