Assay Details
Gene Symbol: Wasf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031877.3 | NP_114083.1 | 7 - 8 | 992 | 69 | |
GenBank mRNA | AF290877.1 | - | 6 - 7 | 888 | 69 | |
AF467773.1 | - | 6 - 7 | 888 | 69 | ||
BC016896.1 | - | 7 - 8 | 950 | 4239038 | 69 | |
BC053423.1 | - | 7 - 8 | 953 | 6509953 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
