Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001136055.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001271430.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001271431.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001271432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001271461.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_001271462.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
NM_007656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034890.mm.1 |
TC0200004159.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001136055.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001271430.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001271431.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001271432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001271461.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_001271462.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
NM_007656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034891.mm.1 |
TC0200004159.mm.1 |
MTA TC |