| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400019444.mm.1 |
NM_001038610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019444.mm.1 |
NM_007826.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019444.mm.1 |
TC1400002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400019445.mm.1 |
NM_001038610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019445.mm.1 |
NM_007826.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019445.mm.1 |
TC1400002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400019446.mm.1 |
NM_001038610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019446.mm.1 |
NM_007826.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019446.mm.1 |
TC1400002578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400019447.mm.1 |
NM_001038610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019447.mm.1 |
NM_007826.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400019447.mm.1 |
TC1400002578.mm.1 |
MTA TC |