Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007826.3 | NP_031852.1 | 4 - 5 | 1721 | 83 | |
| GenBank mRNA | AF090436.1 | - | 4 - 5 | 1265 | 83 | |
| AF102547.1 | - | 4 - 5 | 1265 | 83 | ||
| AJ005669.1 | - | 4 - 5 | 783 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dach1
Genomic Map