Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038610.2 | NP_001033699.1 | 8 - 9 | 2304 | 85 | |
| NM_007826.3 | NP_031852.1 | 9 - 10 | 2460 | 85 | ||
| GenBank mRNA | AF090436.1 | - | 9 - 10 | 2004 | 85 | |
| AF090437.1 | - | 8 - 9 | 1848 | 85 | ||
| AF102547.1 | - | 9 - 10 | 2004 | 85 | ||
| AF129510.1 | - | 8 - 9 | 2066 | 85 | ||
| AJ005669.1 | - | 9 - 10 | 1522 | 85 | ||
| AK047409.1 | - | 8 - 9 | 1170 | 85 | ||
| AK053594.1 | - | 8 - 9 | 1513 | 85 | ||
| AK139060.1 | - | 8 - 9 | 2305 | 85 | ||
| BC059934.1 | - | 8 - 9 | 2082 | 85 | ||
| BC078644.1 | - | 8 - 9 | 1838 | 85 | ||
| BC141130.1 | - | 8 - 9 | 1910 | 9055742 | 85 | |
| BC145440.1 | - | 8 - 9 | 1911 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dach1
Genomic Map