Assay Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007837.4 | NP_031863.3 | 2 - 3 | 142 | 82 | |
GenBank mRNA | AK028120.1 | - | 2 - 3 | 139 | 82 | |
BC013718.1 | - | 2 - 3 | 137 | 4012433 | 82 | |
BY121546.1 | - | 2 - 3 | 142 | 82 | ||
X67083.1 | - | 1 - 2 | 49 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK028120.1,BC013718.1,NM_007837.4,X67083.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:

Citations (1)
Different intracellular pathomechanisms produce diverse Myelin Protein Zero neuropathies in transgenic mice.
- Authors
- Wrabetz L, D'Antonio M, Pennuto M, Dati G, Tinelli E, Fratta P, Previtali S, Imperiale D, Zielasek J, Toyka K, Avila RL, Kirschner DA, Messing A, Feltri ML, Quattrini A
- Journal
- J Neurosci - Page No 2358 (Wed Feb 22 00:00:00 EST 2006)
- Volume (Year)
- 26 (2006)
- Product Usage
- -
