Assay Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007838.2 | NP_031864.2 | 2 - 3 | 408 | 106 | |
| GenBank mRNA | AK081542.1 | - | 2 - 3 | 300 | 106 | |
| AK084267.1 | - | 2 - 3 | 292 | 106 | ||
| AK148095.1 | - | 2 - 3 | 289 | 106 | ||
| AK150683.1 | - | 2 - 3 | 299 | 106 | ||
| BC068132.1 | - | 2 - 3 | 408 | 6808198 | 106 | |
| D89063.1 | - | 2 - 3 | 410 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345064 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345065 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292372 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159026 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127481 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216075 | D89063.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23345078 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05422095 | - | - |
| Whole Mouse Genome G4122A | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216075 | D89063.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138613 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03906157 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03906165 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03906160 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03906161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345064 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23345066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23345066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03906157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03638728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03824080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03638723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03366007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03765966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03638730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03366009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23345093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509553 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509552 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420179_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416493_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509542 | - | - |
| Mouse Genome 430 2.0 Array | 1416493_at | - | - |
| Mouse Genome 430 2.0 Array | 1420179_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416493_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420179_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13200_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13200_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400014091.mm.1 | NM_007838.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014091.mm.1 | NM_007838.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014091.mm.1 | TC0400001722.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014092.mm.1 | NM_007838.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014092.mm.1 | NM_007838.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014092.mm.1 | TC0400001722.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416493_PM_at | - | - |
| Mouse Expression Set 430 | 1416493_at | - | - |
| Murine Genome U74v2 Set | 101074_at | - | - |
| Murine Genome U74v2 Set | 101074_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416493_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416493_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416493_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416493_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 761885 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2680731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2680731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680731 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241093 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241093 | - | - |
| mouse-6 v1.1 (Illumina) | scl24791.10_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680731 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24791.10_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2941677 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2941677 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24791.10_1-S | - | - |

Ddost
Genomic Map